siRNA Details
| Gene Symbol: Mkln1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045552.1 | - | 3 | 495 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB644004.1 | - | 3 | 491 | Chr. 6 - 31385878 - 31398757 [-] on Build GRCm38 | |

Mkln1os
Genomic Map