siRNA Details
| Gene Symbol: E230001N04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053925.1 | - | 1 | 46 | ||
| AK083527.1 | - | 1 | 69 | |||
| AK153588.1 | - | 1 | 34 | |||
| BC096647.1 | - | 1 | 44 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E230001N04Rik| Gene Symbol: E230001N04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053925.1 | - | 1 | 46 | ||
| AK083527.1 | - | 1 | 69 | |||
| AK153588.1 | - | 1 | 34 | |||
| BC096647.1 | - | 1 | 44 | |||