siRNA Details
| Gene Symbol: Gm36794 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC072617.1 | - | 2 | 407 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA316859.1 | - | 2 | 407 | Chr. 5 - 111942353 - 111944235 [-] on Build GRCm38 | |

Gm36794
Genomic Map