siRNA Details
| Gene Symbol: 5730447C08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017643.1 | - | 1 | 214 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY717423.1 | - | 1 | 212 | Chr. 2 - 14020978 - 14021394 [-] on Build GRCm38 | |

5730447C08Rik
Genomic Map