siRNA Details
| Gene Symbol: 5930427J20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020038.1 | - | 1 | 1354 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB099815.1 | - | 1 | 272 | Chr. 18 - 58091077 - 58091400 [-] on Build GRCm38 | |

5930427J20Rik
Genomic Map