siRNA Details
| Gene Symbol: Gm15927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051804.1 | - | 5 | 1445 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB465310.2 | - | 3 | 309 | Chr. 7 - 3632116 - 3634844 [-] on Build GRCm38 | |

Gm15927
Genomic Map