siRNA Details
| Gene Symbol: Gm15927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051804.1 | - | 3 | 1065 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15927| Gene Symbol: Gm15927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051804.1 | - | 3 | 1065 | ||