siRNA Details
| Gene Symbol: 4833447P13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019531.1 | - | 685 | |||
| AK034771.1 | - | 1 | 690 | |||
| AK042445.1 | - | 567 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4833447P13Rik| Gene Symbol: 4833447P13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019531.1 | - | 685 | |||
| AK034771.1 | - | 1 | 690 | |||
| AK042445.1 | - | 567 | ||||