siRNA Details
| Gene Symbol: Gm38411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040387.1 | - | 699 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB209010.2 | - | 1, 2 | 374 | Chr. 3 - 94368615 - 94370159 [-] on Build GRCm38 | |

Gm38411
Genomic Map