siRNA Details
| Gene Symbol: 1500002J14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005119.1 | - | 1 | 152 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY705314.1 | - | 1 | 153 | Chr. 8 - 85701072 - 85701831 [+] on Build GRCm38 | |

1500002J14Rik
Genomic Map