siRNA Details
| Gene Symbol: 4930563J15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016208.1 | - | 2 | 422 | ||
| AK030005.1 | - | 1 | 2584 | |||
| AK030044.1 | - | 1 | 2622 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930563J15Rik| Gene Symbol: 4930563J15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016208.1 | - | 2 | 422 | ||
| AK030005.1 | - | 1 | 2584 | |||
| AK030044.1 | - | 1 | 2622 | |||