siRNA Details
| Gene Symbol: Gm32089 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089519.1 | - | 2, 3 | 198 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY749862.1 | - | 2, 3 | 195 | Chr. 13 - 94289966 - 94295005 [-] on Build GRCm38 | |

Gm32089
Genomic Map