siRNA Details
| Gene Symbol: Mpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK047380.1 | - | 131 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB646947.1 | - | 1 | 132 | Chr. 6 - 50200161 - 50200776 [+] on Build GRCm38 | |

Mpp6
Genomic Map