siRNA Details
| Gene Symbol: Slc35a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009919.1 | - | 1 | 911 | ||
| AK033430.1 | - | 1 | 3052 | |||
| AK081236.1 | - | 1 | 2602 | |||
| AK150620.1 | - | 1 | 1422 | |||
| AK153354.1 | - | 1 | 1422 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ265161.1 | - | 1 | 100 | Chr. 3 - 116669957 - 116670467 [-] on Build GRCm38 | |

Slc35a3
Genomic Map