siRNA Details
| Gene Symbol: Slc35a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009919.1 | - | 1 | 551 | ||
| AK033430.1 | - | 1 | 2691 | |||
| AK081236.1 | - | 1 | 2241 | |||
| AK150620.1 | - | 1 | 1061 | |||
| AK153354.1 | - | 1 | 1061 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35a3| Gene Symbol: Slc35a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009919.1 | - | 1 | 551 | ||
| AK033430.1 | - | 1 | 2691 | |||
| AK081236.1 | - | 1 | 2241 | |||
| AK150620.1 | - | 1 | 1061 | |||
| AK153354.1 | - | 1 | 1061 | |||