siRNA Details
| Gene Symbol: Slc35a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009919.1 | - | 1 | 871 | ||
| AK033430.1 | - | 1 | 3012 | |||
| AK081236.1 | - | 1 | 2562 | |||
| AK150620.1 | - | 1 | 1382 | |||
| AK153354.1 | - | 1 | 1382 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY709506.1 | - | 1 | 872 | Chr. 3 - 116670287 - 116671275 [-] on Build GRCm38 | |

Slc35a3
Genomic Map