siRNA Details
| Gene Symbol: Ano10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049074.1 | - | 2 | 2098 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ano10| Gene Symbol: Ano10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049074.1 | - | 2 | 2098 | ||