siRNA Details
| Gene Symbol: Gm5112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085200.1 | - | 1496 | |||
| AK085233.1 | - | 1495 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5112| Gene Symbol: Gm5112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085200.1 | - | 1496 | |||
| AK085233.1 | - | 1495 | ||||