siRNA Details
| Gene Symbol: Gm5112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085200.1 | - | 1587 | |||
| AK085233.1 | - | 1586 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AW125892.1 | - | 1 | 52 | Chr. 6 - 122812714 - 122813132 [+] on Build GRCm38 | |

Gm5112
Genomic Map