siRNA Details
| Gene Symbol: Gm3646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086013.1 | - | 1, 2 | 162 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB662345.1 | - | 1, 2 | 162 | Chr. 1 - 39804148 - 39805332 [-] on Build GRCm38 | |

Gm3646
Genomic Map