siRNA Details
| Gene Symbol: Gm35363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK158044.1 | - | 1 | 39 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY752807.1 | - | 1 | 39 | Chr. 7 - 101526462 - 101538330 [-] on Build GRCm38 | |

Gm35363
Genomic Map