siRNA Details
| Gene Symbol: 2210402C17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008799.1 | - | 1 | 316 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY551729.1 | - | 1 | 319 | Chr. 11 - 86581201 - 86581698 [-] on Build GRCm38 | |

2210402C17Rik
Genomic Map