siRNA Details
| Gene Symbol: 2900060N18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013539.1 | - | 1 | 272 | ||
| AK019349.1 | - | 1 | 275 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB844529.1 | - | 1 | 501 | Chr. 11 - 119933103 - 119933757 [-] on Build GRCm38 | |

2900060N18Rik
Genomic Map