siRNA Details
| Gene Symbol: Tcrg-V7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | Z48593.1 | - | 1 | 29 | ||
| Z48594.1 | - | 1 | 31 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | M71214.1 | - | 92 | Chr. - - [-] on Build | ||

Tcrg-V7
Genomic Map