siRNA Details
| Gene Symbol: Tcrg-V7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | Z48593.1 | - | 1 | 198 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | M71214.1 | - | 261 | Chr. - - [-] on Build | ||

Tcrg-V7
Genomic Map