siRNA Details
| Gene Symbol: Gm15759 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028685.1 | - | 3349 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15759| Gene Symbol: Gm15759 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028685.1 | - | 3349 | |||