siRNA Details
| Gene Symbol: Gm20756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK081540.1 | - | 1 | 1215 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB366980.2 | - | 1 | 346 | Chr. 6 - 25955355 - 25958159 [+] on Build GRCm38 | |

Gm20756
Genomic Map