siRNA Details
| Gene Symbol: Igdcc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012957.1 | - | 1 | 739 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CN525319.1 | - | 1 | 461 | Chr. 9 - 65186556 - 65187033 [+] on Build GRCm38 | |

Igdcc3
Genomic Map