siRNA Details
| Gene Symbol: 4933436E23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040455.1 | 9 | 973 | |||
| GenBank mRNA | AK017079.1 | - | 9 | 973 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933436E23Rik| Gene Symbol: 4933436E23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040455.1 | 9 | 973 | |||
| GenBank mRNA | AK017079.1 | - | 9 | 973 | ||