siRNA Details
| Gene Symbol: 4933436E23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040455.1 | 8, 9 | 742 | |||
| GenBank mRNA | AK017079.1 | - | 8, 9 | 742 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716757.1 | - | 8, 9 | 738 | Chr. 1 - 139379456 - 139402670 [+] on Build GRCm38 | |

4933436E23Rik
Genomic Map