siRNA Details
| Gene Symbol: 4933436E23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040455.1 | 3 | 289 | |||
| GenBank mRNA | AK017079.1 | - | 3 | 289 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716757.1 | - | 3 | 285 | Chr. 1 - 139379456 - 139402670 [+] on Build GRCm38 | |

4933436E23Rik
Genomic Map