siRNA Details
| Gene Symbol: Crnde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033641.3 | 6, 7 | 1008 | |||
| GenBank mRNA | AK017076.1 | - | 6, 7 | 1008 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crnde| Gene Symbol: Crnde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033641.3 | 6, 7 | 1008 | |||
| GenBank mRNA | AK017076.1 | - | 6, 7 | 1008 | ||