siRNA Details
| Gene Symbol: Crnde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033641.3 | 3 | 562 | |||
| GenBank mRNA | AK017076.1 | - | 3 | 563 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DT913885.1 | - | 1 | 17 | Chr. 8 - 92326124 - 92346870 [-] on Build GRCm38 | |

Crnde
Genomic Map