siRNA Details
| Gene Symbol: Crnde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033641.3 | 4 | 648 | |||
| GenBank mRNA | AK017076.1 | - | 4 | 649 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DT913885.1 | - | 2 | 103 | Chr. 8 - 92326124 - 92346870 [-] on Build GRCm38 | |

Crnde
Genomic Map