siRNA Details
| Gene Symbol: Crnde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033641.3 | 7 | 1074 | |||
| NR_110452.1 | 4 | 893 | ||||
| GenBank mRNA | AK017076.1 | - | 7 | 1074 | ||
| CX205694.1 | - | 4 | 604 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crnde| Gene Symbol: Crnde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033641.3 | 7 | 1074 | |||
| NR_110452.1 | 4 | 893 | ||||
| GenBank mRNA | AK017076.1 | - | 7 | 1074 | ||
| CX205694.1 | - | 4 | 604 | |||