siRNA Details
| Gene Symbol: Crnde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033641.3 | 2 | 482 | |||
| NR_110452.1 | 2 | 482 | ||||
| GenBank mRNA | AK017076.1 | - | 2 | 483 | ||
| CX205694.1 | - | 2 | 193 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY040958.1 | - | 2 | 232 | Chr. 8 - 92346932 - 92355869 [-] on Build GRCm38 | |

Crnde
Genomic Map