siRNA Details
| Gene Symbol: 2610209C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016706.1 | - | 1 | 1099 | ||
| AK085917.1 | - | 1 | 251 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610209C05Rik| Gene Symbol: 2610209C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016706.1 | - | 1 | 1099 | ||
| AK085917.1 | - | 1 | 251 | |||