siRNA Details
| Gene Symbol: 4930432F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019591.1 | - | 1 | 2769 | ||
| BC016220.1 | - | 1 | 1794 | 4485816 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930432F04Rik| Gene Symbol: 4930432F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019591.1 | - | 1 | 2769 | ||
| BC016220.1 | - | 1 | 1794 | 4485816 | ||