siRNA Details
| Gene Symbol: Gm31763 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110511.1 | 7 | 952 | |||
| GenBank mRNA | AK079018.1 | - | 7 | 953 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV381305.2 | - | 4 | 609 | Chr. 10 - 67036581 - 67041326 [+] on Build GRCm38 | |

Gm31763
Genomic Map