siRNA Details
| Gene Symbol: Gm31763 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110511.1 | 1 | 12 | |||
| GenBank mRNA | AK079018.1 | - | 1 | 13 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF161256.1 | - | 67 | Chr. - - [-] on Build | ||

Gm31763
Genomic Map