siRNA Details
| Gene Symbol: 0610040F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040757.1 | 2, 3 | 174 | |||
| NR_104577.1 | 2, 3 | 165 | ||||
| GenBank mRNA | AK002866.1 | - | 2, 3 | 175 | ||
| AK039720.1 | - | 2, 3 | 166 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

0610040F04Rik| Gene Symbol: 0610040F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040757.1 | 2, 3 | 174 | |||
| NR_104577.1 | 2, 3 | 165 | ||||
| GenBank mRNA | AK002866.1 | - | 2, 3 | 175 | ||
| AK039720.1 | - | 2, 3 | 166 | |||