siRNA Details
| Gene Symbol: 1700058M10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006833.1 | - | 1 | 57 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706818.1 | - | 1 | 53 | Chr. 8 - 22609085 - 22609423 [+] on Build GRCm38 | |

1700058M10Rik
Genomic Map