siRNA Details
Gene Symbol: 1700058M10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006833.1 | - | 1 | 197 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706818.1 | - | 1 | 193 | Chr. 8 - 22609085 - 22609423 [+] on Build GRCm38 |
