siRNA Details
| Gene Symbol: Atp7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033254.1 | - | 1 | 1364 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB068760.2 | - | 1 | 273 | Chr. X - 106028430 - 106029027 [+] on Build GRCm38 | |

Atp7a
Genomic Map