siRNA Details
| Gene Symbol: 1700060C16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045732.1 | 5 | 572 | |||
| GenBank mRNA | AK006838.1 | - | 5 | 572 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700060C16Rik| Gene Symbol: 1700060C16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045732.1 | 5 | 572 | |||
| GenBank mRNA | AK006838.1 | - | 5 | 572 | ||