siRNA Details
| Gene Symbol: Gm15050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK142386.1 | - | 524 | |||
| BB660130.1 | - | 524 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15050| Gene Symbol: Gm15050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK142386.1 | - | 524 | |||
| BB660130.1 | - | 524 | ||||