siRNA Details
| Gene Symbol: Gm11494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085785.1 | - | 4 | 509 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB661926.1 | - | 4 | 505 | Chr. 11 - 90135953 - 90143621 [-] on Build GRCm38 | |

Gm11494
Genomic Map