siRNA Details
| Gene Symbol: 6530409C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018330.1 | - | 2 | 761 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY719233.1 | - | 2 | 759 | Chr. 6 - 28215577 - 28218225 [+] on Build GRCm38 | |

6530409C15Rik
Genomic Map