siRNA Details
| Gene Symbol: Ppp6r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020959.1 | - | 1 | 236 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY726992.1 | - | 1 | 233 | Chr. 15 - 89286536 - 89287080 [-] on Build GRCm38 | |

Ppp6r2
Genomic Map