siRNA Details
| Gene Symbol: C78197 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030785.1 | - | 1 | 780 | ||
| AK037678.1 | - | 2 | 530 | |||
| AK041628.1 | - | 2 | 435 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C78197| Gene Symbol: C78197 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030785.1 | - | 1 | 780 | ||
| AK037678.1 | - | 2 | 530 | |||
| AK041628.1 | - | 2 | 435 | |||